Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QPCT Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $437.00
Specifications
Antigen | QPCT |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551074
|
Invitrogen™
MA551074 |
20 μL |
Each for $195.80
|
|
|||||
FEMA551073
|
Invitrogen™
MA551073 |
200 μL |
Each for $437.00
|
|
|||||
Description
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.Specifications
QPCT | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
5730422A13Rik; EC; GCT; Glutaminyl cyclase; glutaminyl-peptide cyclotransferase; glutaminyl-peptide cyclotransferase (glutaminyl cyclase); glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC; Qpct; RGD1562284; similar to Glutaminyl-peptide cyclotransferase precursor (QC); sQC | |
Qpct | |
IgG1 κ | |
Protein A/G | |
Qpct |
C2 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q16769 | |
25797 | |
Recombinant Human QPCT protein, Val29-Leu361 (Accession #Q16769). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title