Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QPRT Antibody (24C4), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB161215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161215 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB161215 Supplier Novus Biologicals Supplier No. NBP259433UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

QPRT Monoclonal antibody specifically detects QPRT in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen QPRT
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 24C4
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias EC 2.4.2.19, nicotinate-nucleotide pyrophosphorylase (carboxylating), QAPRTase, QPRTasenicotinate-nucleotide pyrophosphorylase [carboxylating], quinolinate phosphoribosyltransferase, Quinolinate phosphoribosyltransferase [decarboxylating]
Host Species Mouse
Immunogen Recombinant human QPRT (1-297aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23475
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.