Learn More
Eisco™ QR Code Glass Volumetric Flask with PE Stopper
Catalog No. 12183028 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-183-028 2000 mL
12-183-035 25 mL
12-183-034 50 mL
12-183-033 100 mL
12-183-032 200 mL
12-183-031 250 mL
12-183-030 500 mL
12-183-029 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 12-183-028 Supplier Eisco™ Supplier No. CH3011QRST
Volumetric Flask, Class A. Made of Borosilicate Glass with a Polyethylene Stopper. QR Code Marking for Calibration Certificate.
  • Scan the QR code printed directly on the body of the flask to retrieve a PDF of the calibration batch certificate
  • Made of high quality, heavy duty, borosilicate 3.3 glass. Can withstand high temperatures and thermal shocks
  • Flask features a single white graduation mark
  • Class A tolerance, Conforms to ISO 1042 standards
  • The Flask has a socket fitted with a chemically-resistant polyethylene stopper
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass (3.3)
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Class Class A
Bottom Shape Flat
Surface Treatment Uncoated