Learn More
QRP™ PolyTuff™ Powder Free Solvent Gloves
Catalog No. 19048654c Shop All Protective Industrial Products Inc. Products
Click to view available options
:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Class M3.5 (Class 100) processed and packaged
Class M3.5 (Class 100) processed and packaged
Compounded and processed without chlorides or DOPs. powder free to eliminate particulate contaminants. PolyTuff formulation provides tough, sheer surface that resists snags, punctures, and abrasions. Provides protection from chlorinated solvents. (See permeation breakthrough chart.) Gloves feature Comfort Curve™ design for long wear without finger and hand fatigue.
Permeation Breakthrough Time in Minutes for PolyTuff powder free Solvent Gloves
Reagent Grade Solvent    20G Series (5 mil)    23G Series (1.5 mil)
Methylene Dianiline (MDA)    30-60    <30d-Limonene    <75    12
Xylene    31    3
Carbon Tetrachloride    33    71,1,1-Trichloroethane    35    5
Freon    65    17
Ethanol    21    3
Isopropyl Alcohol (IPA)    50    12
Methyl Ethyl Ketone (MEK)    2    <1
Ethyl Acetate    2    <1
Dimethyl Formamide (DMF)    4    <1
Note: Applicable specification: ASTM 739.
Specifications
Latex Free Yes
Material Vinyl
Powdered Powder Free
Size Large
Sterility Non-sterile
Model 23GL
Length (English) 12 in.
Length (Metric) 30.5 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.