Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ QS 11
GREENER_CHOICE

Catalog No. 332410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-241-0 10 mg
33-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-241-0 Supplier Tocris Bioscience™ Supplier No. 3324/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

ARFGAP1 inhibitor; modulates Wnt/β-catenin signaling

GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1) inhibitor. Modulates ARF-GTP levels and synergizes with the Wnt/β-catenin signaling pathway to upregulate β-catenin nuclear translocation. Also reduces in vitro migration of metastatic human breast cancer cells.

Chemical Identifiers

CAS 944328-88-5
Molecular Formula C36H33N5O2

Specifications

Quantity 10 mg
Formula Weight 567.68
Purity >98%
Molecular Weight (g/mol) 567.693
Chemical Name or Material QS 11
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.