Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QSK Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

Supplier:  Bethyl Laboratories, Inc A302455AT

Encompass_Preferred

 View more versions of this product

Catalog No. 50-156-2630


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

Inhibits CREB activity by phosphorylating and inhibiting activity of TORCs, the CREB-specific coactivators, like CRTC2/TORC2 and CRTC3/TORC3 in response to cAMP signaling.
Specifications

Specifications

QSK
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q6P4S6, Q9Y2K2
Sik3
Between 900 and 950
20 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot
Unconjugated
SIK3
Serine/threonine-protein kinase SIK3, Salt-inducible kinase 3, SIK-3, Serine/threonine-protein kinase QSK, SIK3, KIAA0999, QSK
Rabbit
Antigen affinity chromatography
RUO
23387, 70661
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.