Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QSK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$155.13
Specifications
Antigen | QSK |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2630
![]() |
Bethyl Laboratories, Inc
A302455AT |
20 μL |
Each of 1 for $155.13
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Inhibits CREB activity by phosphorylating and inhibiting activity of TORCs, the CREB-specific coactivators, like CRTC2/TORC2 and CRTC3/TORC3 in response to cAMP signaling.Specifications
QSK | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Serine/threonine-protein kinase SIK3, Salt-inducible kinase 3, SIK-3, Serine/threonine-protein kinase QSK, SIK3, KIAA0999, QSK | |
Between 900 and 950 | |
Primary | |
4° C | |
SIK3 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6P4S6, Q9Y2K2 | |
23387, 70661 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title