Learn More
Applied Biosystems™ QuantStudio™ 7 Flex Real-Time PCR System, 96-well, desktop
Description
Includes
Desktop Computer
With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.
- Increased application versatility through the accommodation of 4 different block types
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, automation capabilities, and effortless block exchange without the need for any tools
Thermo Fisher Cloud:
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.
The QuantStudio™ 7 Flex system accommodates the interchange of a 96-well, 96-well Fast, 384-well, or TaqMan™ array card block. The system is delivered with one block type (a 96-well block in this case); other block types may be purchased separately. In addition, a Twister™ Robot Automation Accessory is available for this system to support high throughput workflows. For a more value-priced system that does not support TaqMan™ array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio™ 6 Flex system. And for the ultimate in flexibility, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan™ OpenArray™ plates.
Accomplish More, Faster:
Run hundreds of real-time PCR reactions effortlessly using TaqMan™ Array 384-well microfluidic cards. And when integrated with the Twister™ Robot Automation Accessory (available separately), the QuantStudio™ 7 Flex system maximizes your throughput capabilities.
High Application Versatility:
Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, High Resolution Melt (HRM), and Protein Thermal Shift™ experiments.
Results You Can Trust:
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex™ system, which features 6 decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
Sensitive Data Analysis:
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Block Format | Interchangeable |
| Format | 96-well Plate |
| Content And Storage | Desktop Computer |
| For Use With (Equipment) | QuantStudio™ |
| Product Line | QuantStudio™ |
| Display Type | Touchscreen |
| Quantity | 1 system |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.