Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qubit™ Flex Starter Packages

Catalog No. p-7235448
Encompass_Preferred
Click to view available options
Type:
Endotoxin Detect. Starter Kit
NGS Starter Kit
Quantitation Starter Kit

Measure fluorescence simultaneously, and reduce assay variability, with the Qubit Flex NGS and Flex Quantitation Starter Kits, which offer highly accurate quantification of DNA, RNA, microRNA, and protein using the Qubit Flex Fluorometer .

Invitrogen Qubit Flex starter kits combine a Qubit Flex Fluorometer with the reagents, plastics, accessories, and training necessary to get starting using the Qubit Flex Fluorometer for quantitation, next-generation sequencing, and endotoxin detection.

The Qubit Flex Fluorometer is a benchtop fluorometer designed for highly accurate quantification of DNA, RNA, microRNA, protein, and endotoxin. With the Qubit Flex Fluorometer you have the flexibility to directly measure fluorescence of up to eight samples simultaneously, which reduces assay variability. Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results.

Qubit Flex Quantitation Starter Kit
(Cat. No. Q45894) includes:
• Qubit Flex Fluorometer (Cat. No. Q33326)
• Qubit Flex Assay Tube Strips, 125 strips (Cat. No. Q33252)
• Qubit 1X dsDNA HS Assay Kit, 100 assays (Cat. No. Q33230)
• Qubit RNA HS Assay Kit, 100 assays (Cat. No. Q32852)
• Qubit RNA BR Assay Kit, 100 assays (Cat. No. Q10210)
• Qubit Protein Assay Kit, 100 assays (Cat. No. Q33211)
• Qubit Flex System Verification Assay Kit (Cat. No. Q33254)
• Qubit Flex Reservoirs, 100 reservoirs (Cat. No. Q33253)
• WiFi Dongle (Cat. No. A26774)
• Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
• Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement

Qubit Flex NGS (Next-Generation Sequencing) Starter Kit (Cat. No. Q45893) includes:
• Qubit Flex Fluorometer (Cat. No. Q33326)
• Qubit Flex Assay Tubes Strips, 125 strips (Cat. No. Q33252)
• Qubit 1X dsDNA HS Assay Kit, 500 assays (Cat. No. Q33231)
• Qubit Flex Assay Reservoirs, 100 reservoirs (Cat. No. Q33253)
• WiFi Dongle (Cat. No. A26774)
• Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
• Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement

Qubit Flex Endotoxin Detection Starter Kit (Cat. No. Q32894) includes:
• Qubit Flex Fluorometer (Cat. No. Q33327)
• Qubit Flex Pyrogen Free Assay Tubes Strips, 120 strips (Cat. No. Q32893)
• Qubit Endotoxin Detection Assay Kit, 80 assays (Cat. No. Q32891)
• Qubit Flex System Verification Assay Kit (Cat. No. Q33254)
• WiFi Dongle (Cat. No. A26774)
• Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
• Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement

TRUSTED_SUSTAINABILITY

Specifications

Sample Volume 1 to 20 μL
Includes Qubit™ Flex Fluorometer, Qubit™ Flex Power Cord, Wi-Fi Dongle, Qubit™ 1X dsDNA HS Assay Kit, Qubit™ Flex Assay Tube Strips
Excitation Wavelength 460 to 480 nm (Blue LED), 620 to 640 nm (Red LED)
Product Line Qubit
Type NGS Starter Kit
For Use With (Application) DNA Quantification, RNA Quantification, MicroRNA Quantification, Protein Quantification
Quantity 1 kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.