Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Quin C1
GREENER_CHOICE

Catalog No. p-7110957
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective FPR2 agonist

Quin C1 is a potent and selective FPR2 agonist (EC50 = 15 nM). Induces Ca2+ mobilization in FPR2 but not FPR1-transfected mast cells. Induces neutrophil chemotaxis and degranulation in vitro. Reduces neutrophil and lymphocyte counts in bronchoalveolar lavage fluid in a mouse lung injury model. Anti-inflammatory.

Specifications

Chemical Name or Material 4-Butoxy-N-[1,4-dihydro-2-(4-methoxyphenyl)-4-oxo-3(2H)-quinazolinyl]benzamide
CAS 786706-21-6
Quantity 10 mg
Target Formyl Peptide Receptor Agonists
Molecular Formula C26H27N3O4
Formula Weight 445.51
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.