Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinidine, 98%, anhydrous

Catalog No. p-3779545
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 56-54-2 | C20H24N2O2 | 324.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00135581,MFCD00198096
InChI Key LOUPRKONTZGTKE-LHHVKLHASA-N
Synonym quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
PubChem CID 441074
ChEBI CHEBI:28593
SMILES COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1

Specifications

Melting Point 170.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 8176
Specific Rotation + 256.00
Solubility Information Solubility in water: 1g/2000mL (20°C)- 1g/800mL (100°C). Other solubilities: 1g/36mL alcohol- soluble in methanol,1g/56mL ether- 1g/1.6mL chloroform,practically insoluble in petroleum ether
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
Formula Weight 324.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Quinidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/at
EINECSNumber 200-279-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.