Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ R 121919 Hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
R 121919 hydrochloride is a high affinity corticotropin releasing factor receptor1 (CRF1) antagonist (Ki = 3.5 nM). Reduces stress-induced elevations of plasma ACTH. Anxiolytic and antidepressant in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-[6-(Dimethylamino)-4-methyl-3-pyridinyl]-2,5-dimethyl-N,N-dipropylpyrazolo[1,5-a]pyrimidin-7-amine hydrochloride |
| CAS | 195055-66-4 |
| Quantity | 10 mg |
| Molecular Formula | C22H32N6.HCl |
| Formula Weight | 416.99 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction