Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog No. p-7054912
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6335103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6335103 Supplier Thermo Scientific Chemicals Supplier No. H6335103

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is used in the preparation of fluorinated analogs of glutemic and glutamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is used in the preparation of fluorinated analogs of glutemic and glutamine.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95715-87-0
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00674064
InChI Key PNJXYVJNOCLJLJ-QMMMGPOBSA-N
Synonym r-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, unii-upk623lv60, r-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, tert-butyl 4r-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, r-+-3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl r-+-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4r, r-garner aldehyde, r-3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde
PubChem CID 178792
IUPAC Name tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
SMILES CC(C)(C)OC(=O)N1[C@H](COC1(C)C)C=O

Specifications

Color Colorless to Yellow
Density 1.06 g/mL
Boiling Point 67°C (0.3 mmHg)
Flash Point 102°C (216°F)
Refractive Index 1.445
Quantity 1 g
Beilstein 3651554
Sensitivity Air sensitive
Merck Index 14,4371
Formula Weight 229.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.