Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Hydroxy-3-phenylpropanoic Acid 99%, Thermo Scientific™

Catalog No. AC412112500
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2768-42-5
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00145219
InChI Key AYOLELPCNDVZKZ-MRVPVSSYSA-M
Synonym 3r-3-hydroxy-3-phenylpropanoic acid, r-3-hydroxy-3-phenylpropanoic acid, r-3-hydroxy-3-phenylpropionic acid, r-+-3-hydroxy-3-phenylpropionic acid, +-3-phenylhydracrylic acid, benzenepropanoic acid, b-hydroxy-, br, r-+-3-hydroxy-3-phenylpropanoic acid, benzenepropanoic acid,, a-hydroxy-,, ar, r-+-3-hydroxy-3-phenyl propionic acid, unii-s2w4lws09c component
PubChem CID 6950815
ChEBI CHEBI:51059
IUPAC Name (3R)-3-hydroxy-3-phenylpropanoate
SMILES O[C@H](CC([O-])=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 116°C to 119°C
Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 250 mg
Specific Rotation Condition +22.50° (20°C c=4,CH3OH)
Specific Rotation +21.5° to +23.5° (20°C, 589nm) (c=4, CH3OH)
Formula Weight 166.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Needles
Chemical Name or Material (R)-3-Hydroxy-3-phenylpropionic acid, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.