Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ R 7050

Click to view available options
Quantity:
10 mg
50 mg
Description
R 7050 is an inhibitor of TNF-α receptor 1 (TNFR1) signaling; blocks TNFR1 association with TRADD and RIP1. Inhibits TNF-α-induced NF-κB and MAPK signaling pathway activation. Attenuates intracerebral hemorrhage-induced neurovascular injury in mice.
Specifications
Specifications
Quantity | 50 mg |
Product Type | R 7050 |
Purity | >98% |
Molecular Weight (g/mol) | 380.77 |
Inhibitors | R 7050 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction