Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Aldosterone Parameter Assay Kit

Catalog No. kge016
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
KGE016 1 Kit
1 options
This item is not returnable. View return policy

Catalog No. KGE016

Supplier: R&D Systems™ KGE016

This item is not returnable. View return policy

Aldosterone Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Aldosterone Parameter Assay Kit quantifies multi-species Aldosterone in cell culture supernates (34 ul), serum (34 ul), edta plasma (34 ul), heparin plasma (34 ul), urine (10 ul).

Specifications

Quantity 1 Kit
Conjugate Biotin
Assay Sensitivity 22.4 pg/mL
Assay Range 24.70 to 6000 pg/mL
For Use With (Application) ELISA
Sample Type Cell Culture Supernates (34uL), Serum (34uL), EDTA Plasma (34uL), Heparin Plasma (34uL), Urine (10uL)
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.