Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-alpha-Methylaspartic acid, 98%, 98% ee, Thermo Scientific™

Catalog No. AC290145000
Click to view available options
Quantity:
250 mg
500 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14603-76-0
Molecular Weight (g/mol) 147.13
InChI Key CWAYDJFPMMUKOI-RXMQYKEDSA-N
Synonym 2-methyl-d-aspartic acid, alpha-methyl-d-asp, r---2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl, 2r-2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl-, hydrochloride 1:1, ambotzhaa7540, a-methyl-d-asp, h-a-me-d-asp-oh, r-2-methylaspartic acid
PubChem CID 6993662
IUPAC Name (2R)-2-amino-2-methylbutanedioic acid
SMILES CC(CC(=O)O)(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >249°C
Color White
Enantiomeric Excess > 75%
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Molecular Formula C5H9NO4
Linear Formula HOOCC(CH3)NH2CH2COOH
Quantity 500 mg
Specific Rotation -54
Formula Weight 147.13
Percent Purity 98%,98% ee
Physical Form Fine Crystalline Powder
Chemical Name or Material (R)-α-Methylaspartic acid, 99+%, (80% e.e.)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.