Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (R)-Citalopram oxalate

Click to view available options
Quantity:
10 mg
50 mg
Description
(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.
Racemate and S-enantiomer also available.
Specifications
Specifications
Chemical Name or Material | (-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate |
CAS | 219861-53-7 |
Quantity | 10 mg |
Target | 5-HT Transporter Inhibitors |
Molecular Formula | C20H21FN2O.C2H2O4 |
Formula Weight | 414.43 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction