Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (R)-CR8
GREENER_CHOICE

Catalog No. p-5545637
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk1 and cdk2; also inhibits CK1; acts as molecular glue

(R)-CR8 is a cyclin dependent kinase (cdk) inhibitor (reported IC50 values are 0.036 - 0.07, 0.09 - 0.8, 0.13 - 0.68, 0.18 - 1 and >1 μM for cdk2, cdk1, cdk5, cdk9 and cdk7, respectively). Also inhibits casein kinase 1 (CK1; IC50 = 0.6 μM). Acts as molecular glue; forms a complex between CDK12-cyclin K and the CUL4 adapter protein DDB1 resulting in ubiquination and degradation of cyclin K.

Specifications

CAS 1786438-30-9
Quantity 10 mg
Molecular Formula C24H29N7O0.3 HCl
Product Type (2R)-2-[[9-(Methylethyl)-6-[[[4-(2-pyridinyl)phenyl]methyl]amino]-9H-purin-2-yl]amino]-1-butanol trihydrochloride
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 540.92
Inhibitors (R)-CR8

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.