Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ C16
GREENER_CHOICE

Catalog No. p-7082189
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PKR inhibitor

C16 is a PKR inhibitor (IC50 = 210 nM). Rescues PKR-dependent translation block in vitro. Decreases Aβ42-induced inflammatory cytokine release and apoptosis in neuronal cultures. Prevents neuroinflammation and neuronal loss in an acute excitotoxic rat model. Brain penetrant.

Specifications

Chemical Name or Material C16
CAS 608512-97-6
Synonym 6,8-Dihydro-8-(1H-imidazol-5-ylmethylene)-7H-pyrrolo[2,3-g]benzothiazol-7-one
Quantity 10 mg
Solubility Soluble to 50mM in DMSO and to 20mM in ethanol
Molecular Formula C13H8N4OS
Formula Weight 268.29
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.