Learn More
R&D Systems™ CellXVivo Human B Cell Expansion Kit
GSA_VASDP
Catalog No. CDK005 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
CDK005 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. CDK005 Supplier R&D Systems™ Supplier No. CDK005
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
  • The following optimized reagents for the expansion of human B cell populations: B Cell Expander Reagents, Reconstitution Buffers
  • Provides enough reagents for the 3-5 fold expansion of a starting population of 107 human B cells.
For the proliferative expansion of human B cell populations.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit