Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KRIBB11
GSA_VA

Catalog No. p-7082195
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

HSF1 inhibitor

KRIBB11 is a heat shock factor (HSF) inhibitor (IC50 = 1.2 μM). Increases apoptosis in cancer cells treated with the HSP90 inhibitors Geldanamycin (Cat. No. 1368) and 17-AAG (Cat. No. 1515). Blocks downstream induction of HSP27 and HSP70. Inhibits tumor growth in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material KRIBB11
CAS 342639-96-7
Synonym N2-1H-Indazol-5-yl-N6-methyl-3-nitro-2,6-pyridinediamine
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C13H12N6O2
Formula Weight 284.27
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.