Learn More
R&D Systems™ MagCellect Mouse Hematopoietic Cell Lineage Depletion Kit
Catalog No. magm209
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Designed to isolate hematopoietic non-lineage committed cells (lineage-negative) from mouse bone marrow
Resulting cell population is highly enriched for CD117+ cells (from 40-70% depending on mouse strain) with less than 5% residual lineage positive cells
Isolated HSCs are untouched by beads or antibodies
Negative selection reduces sources of experimental variation
Lineage committed cells (lineage positive) targeted for magnetic depletion include T cells, B cells, NK cells, monocytes/macrophages, granulocytes, and erythrocytes
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.