Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 179

Click to view available options
Quantity:
10 mg
50 mg
Description
ML 179 is a selective liver receptor homolog 1 (LRH1, NR5A2) inverse agonist (IC50 = 320 nM and maximum efficacy of 64% repression). Exhibits >31-fold selectivity over SF1.
Specifications
Specifications
| Chemical Name or Material | ML 179 |
| CAS | 1883548-87-5 |
| Synonym | 3-Cyclohexyl-6-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]-1H-pyrimidine-2,4-dione |
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in DMSO |
| Molecular Formula | C21H25F3N4O2 |
| Formula Weight | 422.44 |
| Physical Form | Crystalline Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >99% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction