Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CD3+ T Cell Enrichment Column, Mini

Catalog No. MTCC500
Click to view available options
Quantity:
10/Pk.
25/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to prepare purified mouse T cell populations via high affinity negative selection

CD3+ is a universal marker for T cells.

Total CD3+ cell recovery from all available CD3+ cells loaded range between 43% and 51%
Purity of recovered cells ranges between 83% and 90%
Based upon gravitational flow
Simple procedure
No specialized equipment required

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Includes Murine T Cell Selection Columns, 25/Pk; 10X Column Buffer Concentrate
Quantity 10/Pk.
Type Cell Enrichment Column
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.