Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse CD3+ T Cell Enrichment Column, Mini
Click to view available options
Quantity:
10/Pk.
25/Pk.
Description
CD3+ is a universal marker for T cells.
Total CD3+ cell recovery from all available CD3+ cells loaded range between 43% and 51%
Purity of recovered cells ranges between 83% and 90%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Specifications
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Includes | Murine T Cell Selection Columns, 25/Pk; 10X Column Buffer Concentrate |
Quantity | 10/Pk. |
Type | Cell Enrichment Column |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction