Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse CD3+ T Cell Enrichment Column, Small
Designed to prepare purified murine T cell populations via high affinity negative selection
Specifications
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
---|---|
Includes | Murine T Cell Selection Columns, 10X Column Buffer Concentrate |
Type | Cell Enrichment Column |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MTCC5
|
R&D Systems™
MTCC5 |
5/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
MTCC10
|
R&D Systems™
MTCC10 |
10/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
MTCC25
|
R&D Systems™
MTCC25 |
25/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
CD3+ is a universal marker for T cells.
Total T cell recovery from these columns ranged between 45% and 65%
Purity (CD3+ cells) of recovered cells ranged between 81% and 88%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
Murine T Cell Selection Columns, 10X Column Buffer Concentrate | |
Cell Enrichment Column |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title