Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ OICR 9429

Click to view available options
Quantity:
10 mg
50 mg
Description
OICR 9429 is a high affinity WDR5 antagonist (Kd = 93 nM; IC50 = 64 nM); disrupts WDR5-MLL interaction. Exhibits selectivity for WDR5 over 22 methyltransferases and a panel of kinases, GPCRs, ion channels, and transporters. Reduces viability of acute myeloid leukemia cells in vitro. Also disrupts MLL1-RbBP5 interaction.
To request the negative control for OICR 9429, please fill out the OICR 0547 request form on the SGC website.
Specifications
Specifications
| Chemical Name or Material | OICR 9429 |
| Synonym | N-[2-(4-Methylpiperazin-1-yl)-5-[3-(morpholin-4-ylmethyl)phenyl]phenyl]-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide |
| Quantity | 50 mg |
| Solubility | Soluble to 50mM in DMSO |
| Target | Additional Lysine Methyltransferase Compounds |
| Molecular Formula | C29H32F3N5O3 |
| Formula Weight | 555.59 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction