Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Pan Caspase Inhibitor Z-VAD-FMK
GSA_VA

Catalog No. fmk001
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as cell-permeable, irreversible inhibitor of caspase activity. R&D Systems Pan Caspase Inhibitor Z-VAD-FMK binds to the active site of activated proteases irreversibly and inhibits apoptosis.

  • Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying caspase activity
  • Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
  • Regulated by a variety of stimuli, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 mg
Product Type Pan Caspase
Molecular Weight (g/mol) 467Da
Inhibitors Z-VAD-FMK
Form Lyophilized

For research use only. Not for use in diagnostic procedures

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.