Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PFI 4

Catalog No. p-7082201
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective BRPF1 bromodomain inhibitor

PFI 4 is a potent and selective BRPF1B bromodomain inhibitor (IC50 = 172 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of other bromodomains including BRPF2 (BRD1), BRPF3 and BRD4. Disrupts BRPF1 binding to histone H3.3. Cell permeable. Inhibits osteoclastogenesis.

Specifications

Chemical Name or Material PFI 4
CAS 900305-37-5
Synonym N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-pyrrolidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide
Quantity 50 mg
Solubility Soluble to 50mM in DMSO and to 10mM in ethanol
Molecular Formula C21H24N4O3
Formula Weight 380.44
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.