Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PR 619

Click to view available options
Quantity:
10 mg
50 mg
Description
PR 619 is a broad spectrum, reversible DUB inhibitor. Exhibits limited activity against other proteases. Induces accumulation of polyubiquitinated proteins, but has no direct inhibitory effect on the proteasome. Cytotoxic in HEK293T and colorectal cancer cells. Stabilizes microtubule network in oligodendroglial cells.
Specifications
Specifications
| Chemical Name or Material | PR 619 |
| CAS | 2645-32-1 |
| Synonym | 2,6-Diamino-3,5-dithiocyanopyridine |
| Quantity | 50 mg |
| Solubility | Soluble to 10mM in DMSO |
| Molecular Formula | C7H5N5S2 |
| Formula Weight | 223.28 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction