Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 1138452 Hydrochloride

Catalog No. p-7082162
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective prostacyclin IP receptor antagonist

Ro 1138452 hydrochloride is a selective prostacyclin IP receptor antagonist (pKi = 8.3). Exhibits no affinity at other prostanoid receptors (EP1-4, FP and TP) in a radioligand binding assay. Demonstrates analgesic activity in rats. Orally bioavailable.

Specifications

Chemical Name or Material Ro 1138452 hydrochloride
Synonym 4,5-Dihydro-N-[4-[[4-(1-methylethoxy)phenyl]methyl]phenyl]-1H-imidazol-2-amine hydrochloride
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C19H23N3O·HCl
Formula Weight 345.87
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.