Learn More
R&D Systems™ SUMO3 Conjugation Kit
Catalog No. k720
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Sufficient for 5 to 10 x 20uL reactions
Human Small Ubiquitin-like Modifier 3 (SUMO3), also known as SMT3A, is synthesized as a 103 amino acid (aa), propeptide with a predicted 11.5 kDa. SUMO3 contains a two aa C-terminal prosegment. Human SUMO3 shares 83% sequence identity with mouse SUMO3. SUMO3 also has high aa sequence homology to SUMO2 and SUMO4, 87% and 75%, respectively. SUMO3 shares only 47% sequence identity with SUMO1. SUMOs are a family of small, related proteins that can be enzymatically attached to a target protein by a post-translational modification process termed SUMOylation. All SUMO proteins share a conserved Ubiquitin domain and a C-terminal diglycine cleavage/attachment site. Following prosegment cleavage, the C-terminal glycine residue of SUMO3 is enzymatically attached to a lysine residue on a target protein. In humans, SUMO3 is conjugated to a variety of molecules in the presence of the SAE1/UBA2 SUMO-activating (E1) enzyme and the UBE2I/Ubc9 SUMO-conjugating (E2) enzyme. In yeast, the SUMO-activating (E1) enzyme is Aos1/Uba2p. Because of the high level of sequence homology most studies report effects of SUMO2/3. For example, addition of SUMO2/3 was shown to modulate the function of ARHGAP21, a RhoGAP protein known to be involved in cell migration. Other reports indicate that the conjugation by SUMO2/3, but not SUMO1, may represent an important mechanism to protect neurons during episodes of cerebral ischemia. However, studies suggest that SUMO2/3 expression is regulated in an isoform-specific manner since oxidative stress downregulated the transcription of SUMO3 but not SUMO2.
Specifications
Content And Storage Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.