Learn More
R&D Systems™ TACS Blue Label Substrate/Blue Streptavidin-HRP Diluent Kit
Catalog No. 480030bl
Click to view available options
:
30 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
For in situ detection of apoptosis in tissue sections and cells.
TACS™ 2 TdT-Blue Label In Situ Apoptosis Detection Kit is designed for the detection of apoptosis in tissues and cells in culture. All the reagents needed for the detection of DNA fragmentation in situ are provided and this kit can be supplemented with additional reagents for double labeling using immunocytochemistry for antigen detection. In addition, the TACS™ 2 TdT-Blue Label In Situ Apoptosis Detection Kit comes complete with Cytonin™, a detergent-based buffer optimized for the permeabilization of cells prior to labeling, and Nuclear Fast Red for excellent contrast with TACS-Blue LabelTM.
Specifications
Quantity 30 Tests
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.