Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R-Spondin 1 Antibody (OTI11A9), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273948C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273948C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273948C Supplier Novus Biologicals Supplier No. NBP273948C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

R-Spondin 1 Monoclonal specifically detects R-Spondin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen R-Spondin 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI11A9
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Gene Symbols RSPO1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 21-263 of human R-Spondin 1 (NP_001033722) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 284654.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.