Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R-Spondin 3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017160 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017160 Supplier Novus Biologicals Supplier No. NBP281849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

R-Spondin 3 Polyclonal specifically detects R-Spondin 3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen R-Spondin 3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 5-20 ug/mL
Formulation PBS
Gene Alias CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Gene Symbols RSPO3
Host Species Rabbit
Immunogen R-Spondin 3 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human R-Spondin 3. The immunogen is located within amino acids 200 - 250 of R-Spondin 3. Amino Acid Squence: KGERGKKGRERKRKKPNKG
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84870
Test Specificity R-Spondin 3 antibody is human and mouse reactive. At least three isoforms of R-Spondin 3 are known to exist; this antibody will detect all three.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.