Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R-Spondin 3 Antibody (OTI3D5), FITC, Novus Biologicals™
SDP

Catalog No. NBP273949F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273949F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273949F Supplier Novus Biologicals Supplier No. NBP273949F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

R-Spondin 3 Monoclonal specifically detects R-Spondin 3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen R-Spondin 3
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3D5
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Gene Symbols RSPO3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 22-272 of human R-Spondin 3 (NP_116173) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84870.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.