Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Mouse Anti-Mouse SLAMF9, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569819
Description
SLAMF9 Monoclonal antibody specifically detects SLAMF9 in Mouse samples. It is validated for Flow cytometry
Specifications
SLAMF9 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
M349 | |
R718 | |
CD2F-10; Cd2f10; SLAM family member 9; Slamf9 | |
Mouse SLAMF9 Fusion Protein | |
0.1 mg | |
98365 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction