Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Mouse Anti-Mouse SLAMF9, BD Horizon™
Mouse Monoclonal Antibody
$122.00 - $306.00
Specifications
Antigen | SLAMF9 |
---|---|
Clone | M349 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
SLAMF9 Monoclonal antibody specifically detects SLAMF9 in Mouse samples. It is validated for Flow cytometrySpecifications
SLAMF9 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
98365 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
M349 | |
Flow Cytometry | |
R718 | |
Mouse | |
Mouse | |
CD2F-10; Cd2f10; SLAM family member 9; Slamf9 | |
Mouse SLAMF9 Fusion Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title