Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Rabbit Anti-Active Caspase-3, BD Horizon™
Rabbit Monoclonal Antibody
Supplier: BD Biosciences 570311
Description
Active Caspase-3 Monoclonal antibody specifically detects Active Caspase-3 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
Specifications
Active Caspase-3 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Human, Mouse | |
Liquid |
Flow Cytometry | |
C92-605.rMAb | |
R718 | |
Apopain; CASP-3; CASP3; CC3; CPP-32; CPP32; Caspase 3; Yama | |
Human Active Caspase-3 Fragment | |
25 μg | |
836, 12367 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction