Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Rabbit Anti-Active Caspase-3, BD Horizon™
Rabbit Monoclonal Antibody
$188.00 - $468.00
Specifications
Antigen | Active Caspase-3 |
---|---|
Clone | C92-605.rMAb |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Active Caspase-3 Monoclonal antibody specifically detects Active Caspase-3 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
Active Caspase-3 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
836, 12367 | |
IgG κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
C92-605.rMAb | |
Flow Cytometry | |
R718 | |
Rabbit | |
Human, Mouse | |
Apopain; CASP-3; CASP3; CC3; CPP-32; CPP32; Caspase 3; Yama | |
Human Active Caspase-3 Fragment | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title