Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ RA 2

Click to view available options
Quantity:
10 mg
50 mg
Description
RA 2 is a potent negative modulator of calcium-activated potassium channels KCa2.3 and KCa3.1 (IC50 values are 2 and 17 nM, respectively), which displays >55-fold selectively for KCa2 and KCa3.1 over other KCa, Kv and Kir channels. In ex vivo porcine coronary endothelium, RA 2 inhibits SKA 31 (Cat. No. 3670)-induced KCa2/KCa3.1 currents and TRAM 34 (Cat. No. 2946)-insensitive KCa2 currents. Reduces heart rate in mice in a KCa3.1-dependent manner.
Specifications
Specifications
Chemical Name or Material | 1,3-Phenylenebis(methylene)bis(3-fluoro-4-hydroxybenzoate) |
CAS | 1867107-62-7 |
Quantity | 10 mg |
Target | Ca2+-Activated Potassium Channel Modulators |
Molecular Formula | C22H16F2O6 |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction