Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RA 839
GREENER_CHOICE

Catalog No. p-7110881
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Nrf2 activator; inhibits Nrf2/Keap1 interaction

RA 839 is an Nrf2 activator; inhibits Nrf2/Keap1 interaction (Kd = 6 μM). Suppresses LPS-induced iNOS and nitric oxide expression in macrophages. Anti-inflammatory.

Specifications

Chemical Name or Material (3S)-1-[4-[[(2,3,5,6-Tetramethylphenyl)sulfonyl]amino]-1-naphthalenyl]-3-pyrrolidinecarboxylic acid
CAS 1832713-02-6
Quantity 10 mg
Target Nrf2 Modulators
Molecular Formula C25H28N2O4S
Formula Weight 452.57
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.