Learn More
Description
Specifications
Specifications
| Antigen | RAB10 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 400 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Gene Alias | GTP-binding protein RAB10, RAB10, member RAS oncogene family, ras-related GTP-binding protein, ras-related protein Rab-10 |
| Gene Symbols | RAB10 |
| Host Species | Mouse |
| Immunogen | Purified recombinant fragment of human Rab10 expressed in E. Coli. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
