Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17413220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17413220 20 μL
NBP174132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17413220 Supplier Novus Biologicals Supplier No. NBP17413220UL

Rabbit Polyclonal Antibody

RAB10 Polyclonal specifically detects RAB10 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen RAB10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. P61027
Gene Alias GTP-binding protein RAB10, RAB10, member RAS oncogene family, ras-related GTP-binding protein, ras-related protein Rab-10
Gene Symbols RAB10
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the C terminal of Rab10. Immunizing peptide sequence HGIRFFETSAKANINIEKAFLTLAEDILRKTPVKEPNSENVDISSGGGVT.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10890
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.