Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab11A Antibody, Novus Biologicals™
SDP

Catalog No. NB1203612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203612 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203612 Supplier Novus Biologicals Supplier No. NB1203612

Rabbit Polyclonal Antibody

Rab11A Polyclonal specifically detects Rab11A in Human, Porcine, Canine, Primate samples. It is validated for Western Blot.

Specifications

Antigen Rab11A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 6.6 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A
Gene Symbols RAB11A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) G T R D D E Y D Y L F K V V L I C(17) of human Rab11.
Purification Method Affinity Purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8766
Test Specificity NB120-3612 detects Rab 11 from human and non-human primate samples.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.