Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab11A Antibody (STo 7C10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20260A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20260A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20260A32 Supplier Novus Biologicals Supplier No. NBP250260AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab11A Monoclonal specifically detects Rab11A in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Rab11A
Applications Western Blot, ELISA
Classification Monoclonal
Clone STo 7C10
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A
Gene Symbols RAB11A
Host Species Mouse
Immunogen Rab11a peptide KQMSDRRENDM
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8766.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.