Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab11A Antibody (STo 7C10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309677 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309677 Supplier Novus Biologicals Supplier No. NBP250260MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab11A Monoclonal antibody specifically detects Rab11A in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Rab11A
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone STo 7C10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A
Host Species Mouse
Immunogen Rab11A peptide KQMSDRRENDM
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8766
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.