Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB11B Rabbit anti-Human, Alexa Fluor 750, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB080994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080994 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080994 Supplier Novus Biologicals Supplier No. NBP297087AF750

Rabbit Polyclonal Antibody

RAB11B Polyclonal specifically detects RAB11B in Human samples. It is validated for ELISA.

Specifications

Antigen RAB11B
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias GTP-binding protein YPT3, H-YPT3, MGC133246, RAB11B, member of RAS oncogene family, RAB11B, member RAS oncogene family, ras-related protein Rab-11B, YPT3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RAB11B (ABQ59034.1; Gly2-Cys215).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9230
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.