Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB11FIP4 Antibody (OTI13B5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP273764A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273764A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273764A32 Supplier Novus Biologicals Supplier No. NBP273764AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB11FIP4 Monoclonal specifically detects RAB11FIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RAB11FIP4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI13B5
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ARFO2, Arfophilin-2, FIP4-Rab11, FLJ00131, KIAA1821arfophilin-2, MGC11316, MGC126566, RAB11 family interacting protein 4 (class II), rab11 family-interacting protein 4, Rab11-FIP4
Gene Symbols RAB11FIP4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 91-177 of human RAB11FIP4 (NP_116321) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84440.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.